AG/AGCL REFERENCE (100 C) 1 * 1 items
Catalog # JENW68006-39
Supplier: Cole-Parmer
Frequently Bought Together
VWR®, Reinforced Shelves for VWR Ovens and Incubators
Supplier: VWR Collection
Lockings for upright/chest freezer racks
Supplier: GLW
Revco® TDE ULT Upright Freezers, –80 °C
Supplier: REVCO TECHNOLOGIES
Bottles, narrow neck, square or round, with screw top and thread
Supplier: Poulten & Graf
RACK, 1 INCH BELL OPTISEAL TUBES 1 * 1 items
Supplier: Beckman Coulter
O-RING, 8-1/2 OD 1 * 1 items
Supplier: Beckman Coulter
6X6 STD FILTER BAS W/70UM MESH 1 * 1 ST
Supplier: ENTEGRIS
TUBES, PC TK WALL 8X34MM (BOX-100) 1 * 100 items
Supplier: Beckman Coulter
Optima MAX-TL Benchtop Ultracentrifuge
Supplier: Beckman Coulter
Steam Cleaner type ElmaSteam 8 Basic, with 1 x fixed nozzle, 1 x hand pi - Schuko plug 1 * 1 items
Supplier: ELMA SCHMIDBAUER
Silicone encased keyboards with full layout, integrated touch pad and backlit keys, GCQ CleanType® Prime Touch+
Supplier: GETT GERATETECHNIK
Pipetman m p10m bt connected cal.3x4 1 * 1 items
Supplier: GILSON
Adapters for Huber circulators
Supplier: HUBER
Cleanroom tables, Ergo
Supplier: Palbam