0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
SURESTART webseal mat, blue, ptfe/silicone, for 96 square well plates, 8mm, flat base, large penetration area, pre-slit, 1 * 5 items
HYPE60180-M122
:
Thermo Fisher Scientific
SURESTART webseal mat, blue, ptfe/silicone, for 96 square well plates, 8mm, flat base, large penetration area, pre-slit, 1 * 5 items
HYPE60180-M122
:
Thermo Fisher Scientific
Frequently Bought Together
ISAspher HPLC Columns, C18 AQ
: ISERA
Solid phase extraction cartridges, Bond Elut™ C18
: VARIAN
ISAspher HPLC Columns, SCX
: ISERA
Grounding cables, b.safe
: Bohlender
Capillary columns, OPTIMA® WAX
: MACHEREY-NAGEL
Gas-clean filters
: VARIAN
Transition fittings, BOLA
: Bohlender
Avantor® ACE® HILIC-N, HPLC/UHPLC Columns, Analytical
: Avantor
Manual crimper for 20 mm crimp vials, level 3, SureSTART™
: Thermo Fisher Scientific
ISAspher HPLC Columns, C18-AQ-PolE
: ISERA