ContactUsLinkComponent
SURESTART webseal mat, blue, ptfe/silicone, for 96 round well plates, 8mm diameter, flat base, no slit, large penetration area, 1 * 5 items
Catalog # HYPE60180-M102
SURESTART webseal mat, blue, ptfe/silicone, for 96 round well plates, 8mm diameter, flat base, no slit, large penetration area, 1 * 5 items
Catalog # HYPE60180-M102
Frequently Bought Together