ContactUsLinkComponent
Corning PET Bottle, 30mL, Graduated, 18mm Screw Cap, Sterile, Triple-bagged, Pre-assembled, 24/Tray, 240/Case 1 * 240 items
Catalog # CORN432229
Supplier:  Corning
Corning PET Bottle, 30mL, Graduated, 18mm Screw Cap, Sterile, Triple-bagged, Pre-assembled, 24/Tray, 240/Case 1 * 240 items
Catalog # CORN432229
Supplier:  Corning
Frequently Bought Together