ContactUsLinkComponent
Laboratory bottle 1000ml, GL45, clear glass, without cap, Boro 3.3, with graduation 1 * 1 items
Catalog # NEUB0851-100100045
Laboratory bottle 1000ml, GL45, clear glass, without cap, Boro 3.3, with graduation 1 * 1 items
Catalog # NEUB0851-100100045
Frequently Bought Together