ContactUsLinkComponent
for checking the photometric accuracy, consisting of: - Glass filter 1, NG9/1, our serial no. F1881, measured against the WG305/2. - Glass filter 2, NG5/2, our serial no. F1882, measured against the WG305/2 - Glass filter 3, NG11/2, our serial no. F1 1 * 1 items
Catalog # HELL2X666S002RE
Supplier:  HELLMA
for checking the photometric accuracy, consisting of: - Glass filter 1, NG9/1, our serial no. F1881, measured against the WG305/2. - Glass filter 2, NG5/2, our serial no. F1882, measured against the WG305/2 - Glass filter 3, NG11/2, our serial no. F1 1 * 1 items
Catalog # HELL2X666S002RE
Supplier:  HELLMA
Frequently Bought Together