MANGANESE (II) CHLORIDE TETRAHYDRATE 1 * 100 g
SIAL221279-100G
: Merck
Frequently Bought Together
Potassium iodide ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals