ContactUsLinkComponent
100 assays Proteinase K Assay [Colorimetric] is designed for rapid and reproducible quantification of Proteinase K activity. Proteinase K Assay [Colorimetric] is based on hydrolysis of denatured hemoglobin by proteinase K which releases Folin positiv 1 * 1 KIT
Catalog # GENO786-1807
Supplier:  G-Biosciences
100 assays Proteinase K Assay [Colorimetric] is designed for rapid and reproducible quantification of Proteinase K activity. Proteinase K Assay [Colorimetric] is based on hydrolysis of denatured hemoglobin by proteinase K which releases Folin positiv 1 * 1 KIT
Catalog # GENO786-1807
Supplier:  G-Biosciences
Frequently Bought Together