A zwitterionic detergent 1 * 500 mL
Catalog # ABCAAB291109-500
Supplier: Abcam
Frequently Bought Together
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Trichloroethylene 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific