(R)-(+)-7-Bis(3,5-di-t-butylphenyl)phosphino-7[(pyridine-2-ylmethyl)amino]-2,2,3,3-tetrahydro-1,1-spirobiindane 1 * 100 mg
Catalog # ABCRAB470533
Supplier: abcr
(R)-(+)-7-Bis(3,5-di-t-butylphenyl)phosphino-7[(pyridine-2-ylmethyl)amino]-2,2,3,3-tetrahydro-1,1-spirobiindane 1 * 100 mg
Catalog # ABCRAB470533
Supplier: abcr
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals