ANIONTOVÝ STANDARD HYDROGEN DRASELNÉ SOLI KYSELINY FTALOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICKHP1-125ML
Supplier: RIVOLAB
ANIONTOVÝ STANDARD HYDROGEN DRASELNÉ SOLI KYSELINY FTALOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICKHP1-125ML
Supplier: RIVOLAB
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
Hydrochloric acid 1% Reag. Ph. Eur.
Supplier: VWR Chemicals
Conductivity standards, Potassium chloride, IUPAC
Supplier: Hach
Triazamate 1 * 50 mg
Supplier: EHRENSTORFER
(±)-Camphene
Supplier: Merck
pH buffer solutions, colour coded
Supplier: Hach
Ethyl Acetate, Reference Standard
Supplier: USP
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals