ANIONTOVÝ STANDARD KYSELINY VINNÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
RIVOICTRTR1-125ML
: RIVOLAB
ANIONTOVÝ STANDARD KYSELINY VINNÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
RIVOICTRTR1-125ML
: RIVOLAB
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
Kerosene, 194…250 °C, J.T.Baker®
: Avantor
Ammonia solution 17% Reag. Ph. Eur. 1004701
: VWR Chemicals
pH buffers, ready to use
: WTW
pH buffers, for LAQUAtwin meters
: HORIBA
Ethylene glycol, Supelco®
: Merck