ANIONTOVÝ STANDARD SODNÉ SOLI KYSELINY ŠŤAVELOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICOXA1-125ML
Supplier: RIVOLAB
ANIONTOVÝ STANDARD SODNÉ SOLI KYSELINY ŠŤAVELOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICOXA1-125ML
Supplier: RIVOLAB
Frequently Bought Together
Accessories for pH/°C meter, handheld
Supplier: Testo
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Hydrochloric acid 1% Reag. Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ