ANIONTOVÝ STANDARD SODNÉ SOLI KYSELINY OCTOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICOAC1-125ML
Supplier: RIVOLAB
ANIONTOVÝ STANDARD SODNÉ SOLI KYSELINY OCTOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICOAC1-125ML
Supplier: RIVOLAB
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonia solution 17% Reag. Ph. Eur. 1004701
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
pH buffer, single-use, SINGLET™
Supplier: Hach
Potassium chloride 3 M in aqueous solution
Supplier: Thermo Fisher Scientific