ANIONTOVÝ STANDARD KYSELINY MALONOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICMLO1-125ML
Supplier: RIVOLAB
ANIONTOVÝ STANDARD KYSELINY MALONOVÉ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICMLO1-125ML
Supplier: RIVOLAB
Frequently Bought Together
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
Triethyl Citrate, Reference Standard
Supplier: USP
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
pH Standard Solutions, IUPAC, Certified
Supplier: Hach