ANIONTOVÝ STANDARD SODNÉ SOLI KYSELINY MRAVENČÍ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICHCO1-125ML
Supplier: RIVOLAB
ANIONTOVÝ STANDARD SODNÉ SOLI KYSELINY MRAVENČÍ PRO IONTOVOU CHROMATOGRAFII, KONCENTRACE 1000UG/ML 1 * 125 mL
Catalog # RIVOICHCO1-125ML
Supplier: RIVOLAB
Frequently Bought Together
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions
Supplier: Hanna
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
Supplier: EHRENSTORFER
BRILLIANT BLACK BN 1 * 0,25 g
Supplier: EHRENSTORFER
Aromatic VOC Mixture 881 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Pinoxaden certified 1 * 25 mg
Supplier: EHRENSTORFER
Sulfite lr tablets in pack of 100 1 * 100 Tablet
Supplier: Lovibond
PAH-Mix 14 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
VWR® ARISTAR®, Molybdenum Standard Solutions for ICP-MS
Supplier: VWR Chemicals
TROLAMINE IMPURITY A ETHANOLAMINE 1 * 250 mg
Supplier: EDQM
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Stearyl Alcohol, Reference Standard
Supplier: USP
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea)
Supplier: EHRENSTORFER