HEXACHLOROBENZENE (HCB) 100G/ML IN CYC 1 * 5 mL
Catalog # RIVO159002660100CY
Supplier: RIVOLAB
Frequently Bought Together
Buffer solutions
Supplier: REAGECON
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Osmolality standards
Supplier: VWR Chemicals
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
METHYL LINOLEATE 1 * 5 items
Supplier: Merck
Chloride Standard for IC 1 * 100 mL
Supplier: Merck