ContactUsLinkComponent
Objective N PLAN 100x/1.25 OIL PH3 Free working distance: 0.12 mm For use with Cover Glass Suitable for Phase Contrast Not suitable for Incident Light, except Fluorescence Useful and recommended for Integrated Leica Modulation Contrast (IMC) 1 * 1 items
Catalog # LEIM11506159
Supplier:  LEICA MICROSYSTEMS
Objective N PLAN 100x/1.25 OIL PH3 Free working distance: 0.12 mm For use with Cover Glass Suitable for Phase Contrast Not suitable for Incident Light, except Fluorescence Useful and recommended for Integrated Leica Modulation Contrast (IMC) 1 * 1 items
Catalog # LEIM11506159
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together