MonoRab Anti-Camelid VHH Affinity Resin ml Resin (Total 20 ml) 1 * 1 KIT
GENSL00905-10
Frequently Bought Together
Laboratory trays
: KARTELL
VWR®, Microcentrifuge Tubes and Screw Caps
: VWR Collection
Media bottles, square, sterile, Nalgene®
: Thermo Fisher Scientific
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
Measuring scoops
: VITLAB
Centrifuge tubes
: Brand
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
Lab blender bags, Stomacher®
: Seward (FermionX)
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Sampling Spoons
: VWR Collection