KJELDAHL TABLETS TITANIUM MICRO Advantages:,,Reduces the amount of chemicals required Composition:,,1,5 g K2So4, 0,045 g CuSo4 * 5 H2O, 0,045 g TiO2 1 * 1.000 items
Catalog # BUCH11072628
Supplier: BUCHI
KJELDAHL TABLETS TITANIUM MICRO Advantages:,,Reduces the amount of chemicals required Composition:,,1,5 g K2So4, 0,045 g CuSo4 * 5 H2O, 0,045 g TiO2 1 * 1.000 items
Catalog # BUCH11072628
Supplier: BUCHI
Frequently Bought Together
Visual colour kits, ammonium, MQuant®, Supelco®
Supplier: Merck
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Supelco® Spectroquant® Test Kits, Sulphate
Supplier: Merck
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach