(S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(di (3,5-dimethylphenyl)phosphino)ferrocene 97% 1 * 100 mg
Catalog # ABCRAB170446
Supplier: abcr
(S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(di (3,5-dimethylphenyl)phosphino)ferrocene 97% 1 * 100 mg
Catalog # ABCRAB170446
Supplier: abcr
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals