Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(S,S)-(-)-2,2-Bis[(R)-(N,N-dimethylamino) 1 100 mg 95,50 (phenyl)methyl]-1,1-bis(di(3,5- dimethylphenyl)phosphino)ferrocene 97% 1 * 100 mg
Catalog # ABCRAB249152
Supplier:
abcr
Expand image
(S,S)-(-)-2,2-Bis[(R)-(N,N-dimethylamino) 1 100 mg 95,50 (phenyl)methyl]-1,1-bis(di(3,5- dimethylphenyl)phosphino)ferrocene 97% 1 * 100 mg
Catalog # ABCRAB249152
Supplier:
abcr
Frequently Bought Together
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Petroleum spirit, 80…110 °C, TECHNICAL
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals