SARS CoV 2 Spike protein (S1 T19R G142D del 156 157 R158G L452R T478K D614G P681R His Tag) 1 * 100 µG
Catalog # GENSZ03612-100
Supplier: Genscript
SARS CoV 2 Spike protein (S1 T19R G142D del 156 157 R158G L452R T478K D614G P681R His Tag) 1 * 100 µG
Catalog # GENSZ03612-100
Supplier: Genscript
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Chemical bottles, wide mouth, square
Supplier: RIXIUS
Measuring scoops
Supplier: KARTELL
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
PCR STRIP W.8X0.2ML TUBE FLAT CAP IND. 1 * 126 items
Supplier: RATIOLAB
Filter plates, 24-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)
Cell culture inserts for multiwell plates, ThinCert™
Supplier: Greiner Bio-One
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
Assay microplates and blocks, 384-well, ScreenMates
Supplier: THERMO MATRIX TECHNOLOGIES
Caps for sample tubes, tamper evident
Supplier: Simport Scientific
Crystallisation plates, CrystalStar™, 96-well CrystalQuick™ and CrystalDrop™
Supplier: Greiner Bio-One
BOTTLE PET 350ML RECT STER 18MG WHITE DOME CAP / 1 * 174 items
Supplier: LAB3 OWN PRODUCTION