High Affinity Ni Charged Resin FF Prepacked Column 5 ml x 5 1 * 5 mL
Catalog # GENSL00683-55
Supplier: Genscript
High Affinity Ni Charged Resin FF Prepacked Column 5 ml x 5 1 * 5 mL
Catalog # GENSL00683-55
Supplier: Genscript
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Sampling Spoons
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Spreaders, T-shaped
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cell culture flasks, Corning®
Supplier: Corning