Reagecon Barium Chloride TS/RS Solution according to United States Pharmacopoeia (USP) 1 * 100 mL
REGEUSP1001
: REAGECON
Reagecon Barium Chloride TS/RS Solution according to United States Pharmacopoeia (USP) 1 * 100 mL
REGEUSP1001
: REAGECON
Frequently Bought Together
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
: PanReac AppliChem
4-(Methylamino)phenol sulphate 99%, extra pure
: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
: Honeywell Chemicals
2-Propanol 70 % pure 1 * 1 L
: PanReac AppliChem
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
HCL ACID REAGENT 1 * 20 L
: Hach
Cyclohexane 99.5%, extra pure
: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
: Thermo Fisher Scientific