2-HYDROXY-5-NITROBENZOIC ACID 1 * 100 mg
Catalog # PHR9056-100MG
Supplier: Merck
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion