(R)-1-[(R)-1-[DI-TERT-BUTYLPHOSPHINO]ETHYL]-2-[2-(DI-TERT-BUTYLPHOSPHINO) PHENYL]FERROCENE 1 * 250 mg
Catalog # ABCRAB426511-250
Dodavatel: abcr
(R)-1-[(R)-1-[DI-TERT-BUTYLPHOSPHINO]ETHYL]-2-[2-(DI-TERT-BUTYLPHOSPHINO) PHENYL]FERROCENE 1 * 250 mg
Catalog # ABCRAB426511-250
Dodavatel: abcr
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dodavatel: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Dodavatel: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Dodavatel: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals