(2S,2S,3S,3S)-3,3-Di-tert-butyl-2,2-diethyl-2,2,3,3-tetrahydro-4,4-bibenzo[d][1,3]oxaphosphole, min. 97% (99% ee)(2S,2S,3S,3S)-DI-Et -BABIBOP CAS 2415751-83-4 1 * 100 mg
Catalog # ABCRAB541715-100
Supplier: abcr
(2S,2S,3S,3S)-3,3-Di-tert-butyl-2,2-diethyl-2,2,3,3-tetrahydro-4,4-bibenzo[d][1,3]oxaphosphole, min. 97% (99% ee)(2S,2S,3S,3S)-DI-Et -BABIBOP CAS 2415751-83-4 1 * 100 mg
Catalog # ABCRAB541715-100
Supplier: abcr
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals