DOWSIL 1-2577 CONFORMAL COATING 1 * 199,5 kg
Katalogové číslo DOWC1-2577-440LB
Supplier: DOW CORNING
DOWSIL 1-2577 CONFORMAL COATING 1 * 199,5 kg
Katalogové číslo DOWC1-2577-440LB
Supplier: DOW CORNING
Frequently Bought Together
Benzyl benzoate for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.95% for DNA synthesis
Supplier: VWR Chemicals
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Chloroform, extra dry 99.9% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1,10-Phenanthroline monohydrate, Supelco®
Supplier: Merck
Xylene (mixture of isomers) for histology
Supplier: Honeywell Chemicals
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Dichloromethane ≥99.7% stabilised for HPLC
Supplier: Thermo Fisher Scientific