Disc 2 for autosampler 1 * 1 items
Catalog # VELPA00000199
Supplier: VELP SCIENTIFIC
Frequently Bought Together
PISTON WITH COUPLING PTFE 20 ML 1 * 1 items
Supplier: METROHM
Short-term tubes for accuro®
Supplier: Dräger
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: COLLECTION PREFIX HPLC
VWR®, Accessories for SAS Super DUO 360
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Precolumn filters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Accessories for Mettler balances
Supplier: Mettler - Toledo
Accessories for titrators
Supplier: Mettler - Toledo
Accessories for compact automatic extraction units, Soxtherm
Supplier: GERHARDT
VWR®, Microscope Cleaning Set
Supplier: VWR Collection
Accessories for VWR® UV-6300PC
Supplier: VWR Collection
VWR®, Digital Calipers
Supplier: VWR Collection
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
GLASS CYLINDER FOR 10 ML BURETTE 1 * 1 items
Supplier: Mettler - Toledo
Melting point and boiling point capillaries and tubes
Supplier: BUCHI