ContactUsLinkComponent
Panthera Cloud Binocular head Siedentopf type, 25 inclined, 360 swiveling with built-in digital camera CMOS 1/2.5 Still resolution: 8MP Live resolution: FullHD (1920x1080) Output: HDMI and RJ45 Widefield high eyepoint eyepieces UC-WF10X/22mm wit 1 * 1 items
Catalog # MOTI1100104601181
Supplier:  MOTIC
Panthera Cloud Binocular head Siedentopf type, 25 inclined, 360 swiveling with built-in digital camera CMOS 1/2.5 Still resolution: 8MP Live resolution: FullHD (1920x1080) Output: HDMI and RJ45 Widefield high eyepoint eyepieces UC-WF10X/22mm wit 1 * 1 items
Catalog # MOTI1100104601181
Supplier:  MOTIC
Frequently Bought Together