ContactUsLinkComponent
Masterflex® Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1" ID x 1" ID x 3/4" NPT(M); 100/PK
Masterflex® Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1" ID x 1" ID x 3/4" NPT(M); 100/PK
Catalog # MFLX30627-84
CE certificate
undefined
Masterflex® Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1" ID x 1" ID x 3/4" NPT(M); 100/PK
Catalog # MFLX30627-84
CE certificate
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1" ID x 1" ID x 3/4" NPT(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
  • Barb size/tubing int.Ø (port 2):
    1"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    T
  • Description:
    Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1" ID x 1" ID x 3/4" NPT(M); 100/PK
  • Seq:
    16
  • Environmentally Preferable:
  • Model:
    30627-84
  • Orifice dimensions:
    0,78125"
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    100
  • Fitting material:
    HDPE
Frequently Bought Together