Zadání objednávky
Czech Republic
ContactUsLinkComponent
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 5/16" ID x 3/8" NPT(M); 100/PK
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 5/16" ID x 3/8" NPT(M); 100/PK
Catalog # MFLX30625-58
CE certificate
undefined
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 5/16" ID x 3/8" NPT(M); 100/PK
Catalog # MFLX30625-58
CE certificate
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 5/16" ID x 3/8" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb
  • Velikost ozubu/vnit. Ø hadičky (port 2):
    ⁵⁄₁₆"
  • Typ připojení:
    Thread to Barb
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ⅜"
  • Popis:
    Fitting, HDPE, Straight, Hosebarb to Thread Adapter, ⁵⁄₁₆" ID x ⅜" NPT(M); 100/PK
  • Sekvence:
    45
  • Environmentally Preferable:
  • Model:
    30625-58
  • No. of ports:
    2
  • Sterilizace:
    Ethylene oxide (ETO) only
  • Bal.:
    100
  • Fitting material:
    HDPE
Frequently Bought Together