ContactUsLinkComponent
Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1/2" ID x 3/8" ID; 10/PK
Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1/2" ID x 3/8" ID; 10/PK
Catalog # MFLX30616-24
CE certificate
undefined
Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1/2" ID x 3/8" ID; 10/PK
Catalog # MFLX30616-24
CE certificate
About this item
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/2" ID x 3/8" ID
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Select the fitting based on the Masterflex tubing size. Fittings are available in a variety of materials to suit different applications and budgets.
  • Barb size/tubing int.Ø (port 1):
    0,5"
  • Barb size/tubing int.Ø (port 2):
    0,375"
  • Connection Type:
    Barb
  • Flow pattern:
    Straight
  • Description:
    Kynar REDUCER,I/P 82/88 & BT 88,10/P
  • Seq:
    22
  • Environmentally Preferable:
  • Model:
    30616-24
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    10
  • Fitting material:
    PVDF
Frequently Bought Together