ContactUsLinkComponent
Smart Gasket Sanitary Sampling Gasket, Silicone, 4 Port, 2" Tri-Clamp
Smart Gasket Sanitary Sampling Gasket, Silicone, 4 Port, 2" Tri-Clamp
Catalog # MFLX30545-16
undefined
Smart Gasket Sanitary Sampling Gasket, Silicone, 4 Port, 2" Tri-Clamp
Catalog # MFLX30545-16
About this item
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
  • Sanitary clamp size (Port 1):
    2"
  • Connection type:
    Sanitary
  • Description:
    2" Tri-Clamp, 4 Port, Silicone, Each
  • Seq:
    15
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    Platinum-cured silicone
  • Sanitary gasket size:
    2"
  • No. of ports:
    4
  • Warranty:
    1 year
Frequently Bought Together