Sulphur S - 1000 mg/l in H2O for ICP CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 30 mL
Katalogové číslo ACSDC049.W.L03
Supplier: CPACHEM
Frequently Bought Together