Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
Polypropylene flexible liner pump, rubber liner, 1 GPM, 115/230 VAC
  MFLX79440-00
undefined
Polypropylene flexible liner pump, rubber liner, 1 GPM, 115/230 VAC
  MFLX79440-00

 

Unique sealless design permits gentle pumping action that provides little churning or foaming.

  • Transfer liquids containing up to 20% soft solids
  • Sealless design with secondary containment system prevents potentially hazardous leakage
  • Fixed-speed pumps are self-priming, reversible, and can run dry without damage

Polypropylene pumps have a Nordel® rubber liner; PTFE pumps have a Hypalon® rubber liner. Variable-speed models and DC-powered pumps are available — call our Applications Specialists for more information.

  • Maximální průtok:
    3,8 L/min (1 gal./min)
  • Napájení:
    115/230 VAC; 60 Hz; 3,5 A
  • Popis:
    Polypropylene flexible liner pump, rubber liner, 1 gal./min, 115/230 VAC
  • Výška:
    206,4 mm
  • Délka:
    38,1 cm
  • Environmentally Preferable:
  • Máčené materiály:
    Polypropylene, Nordel®
  • Duty cycle continuous:
    Yes
  • Inlet connection:
    3/4" hose barb
  • Motor hp:
    1/4
  • Outlet connection:
    3/4" hose barb
  • Power conduit box connection:
    Yes
  • Run dry:
    Yes
  • Self-priming:
    Yes
  • Suction lift, dry:
    1800 mm of H₂O (72" H₂O); 629 mm Hg (5,3" Hg)
  • Maximální teplota:
    85 °C (185 °F)
  • Záruka:
    1 year
  • Bal.:
    1
  • Šířka:
    17,78 cm
Frequently Bought Together