ContactUsLinkComponent
Masterflex® Dual-Drive Syringe Pump RS-232 Connector Cable
Masterflex® Dual-Drive Syringe Pump RS-232 Connector Cable
Catalog # MFLX74901-88
undefined
Masterflex® Dual-Drive Syringe Pump RS-232 Connector Cable
Catalog # MFLX74901-88
About this item
Daisy chain up to 100 pumps.
  • Connect multiple independent syringes to operate simultaneously
  • Easily connect pumps
  • Choose cable based on your current pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
  • Description:
    Pumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe Pump
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    74901-88
  • Outlet connection:
    RS-232 Connector Cable for 74901-80
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together