ContactUsLinkComponent
Cadence Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Lock to 1/4-28 UNF
Cadence Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Lock to 1/4-28 UNF
Catalog # MFLX41507-73
undefined
Cadence Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Lock to 1/4-28 UNF
Catalog # MFLX41507-73
About this item
Excellent durability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
  • Full material traceability on all items
Constructed of durable plated brass, these Luer Adapter Fittings are ideal for a wide range of applications.
  • Connection Type:
    Male Luer to Thread
  • Flow pattern:
    Straight
  • Description:
    Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Lock to 1/4-28 UNF
  • Environmentally Preferable:
  • Connection type (port 1):
    Male luer lock
  • Connection type (port 2):
    1/4-28 UNF
  • No. of ports:
    2
  • Pk:
    1
  • Fitting material:
    Nickel-Plated Brass
  • Thread size UNF(M) (port 1):
    1/4"-28
Frequently Bought Together