Cadence Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x Female Luer
MFLX41507-10
Exceptional durability.
- Easy to disassemble for cleaning
- Stopcock handles turn 180°
- Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.
- Flow pattern:Three-Way
- Popis:Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Female Luer x Male Luer Lock x Female Luer; 1/Ea
- Environmentally Preferable:
- Máčené materiály:Nickel-Plated Brass
- Bal.:1
- Inlet port:Female Luer
- Outlet port:Male Luer Lock and Female Luer
Frequently Bought Together