ContactUsLinkComponent
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Male Luer Lock x Male Luer Lock
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Male Luer Lock x Male Luer Lock
Catalog # MFLX41507-02
undefined
Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Male Luer Lock x Male Luer Lock
Catalog # MFLX41507-02
About this item
Exceptional durability.
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.
  • Flow pattern:
    One-Way
  • Description:
    Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Male Luer Lock x Male Luer Lock; 1/Ea
  • Environmentally Preferable:
  • Wetted materials:
    Nickel-Plated Brass
  • Pk:
    1
  • Inlet port:
    Male Luer Lock
  • Outlet port:
    Male Luer Lock
Frequently Bought Together