ContactUsLinkComponent
Threaded Reducing Bushing, 1" NPT(M) x 3/4" NPT(F), PTFE
Threaded Reducing Bushing, 1" NPT(M) x 3/4" NPT(F), PTFE
Catalog # MFLX34030-54
undefined
Threaded Reducing Bushing, 1" NPT(M) x 3/4" NPT(F), PTFE
Catalog # MFLX34030-54
About this item
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings offer more permanent installation. Choose the size and material that meets your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 2):
    ¾"
  • Thread size NPT(M) (port 1):
    1"
  • Description:
    Threaded Reducing Bushing, 1" NPT(M) × ¾" NPT(F), PTFE
  • Seq:
    50
  • Environmentally Preferable:
  • Model:
    34030-54
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    1
  • Fitting material:
    PTFE
Frequently Bought Together