ContactUsLinkComponent
Compression pipe adapter, Brass, 3/4", Each
Compression pipe adapter, Brass, 3/4", Each
Catalog # MFLX31412-80
undefined
Compression pipe adapter, Brass, 3/4", Each
Catalog # MFLX31412-80
About this item
Create a secure connection with compression fittings.
  • Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 2):
    3/4"
  • Description:
    Compression pipe adapter, Brass, 3/4", Each
  • Environmentally Preferable:
  • Warranty:
    LTD life time
  • Pk:
    1
  • Fitting material:
    Brass
Frequently Bought Together