ContactUsLinkComponent
316 SS Female Reducer, 1/4" x 1/8" NPT(F) 31211-10
316 SS Female Reducer, 1/4" x 1/8" NPT(F) 31211-10
Catalog # MFLX31211-10
undefined
316 SS Female Reducer, 1/4" x 1/8" NPT(F) 31211-10
Catalog # MFLX31211-10
About this item
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    ¼"
  • Thread size NPT(F) (port 2):
    ⅛"
  • Description:
    316 SS Female Reducer, ¼" × ⅛" NPT(F) 31211-10
  • Seq:
    07
  • Environmentally Preferable:
  • Model:
    31211-10
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Pk:
    1
  • Fitting material:
    Stainless Steel 316
Frequently Bought Together