ContactUsLinkComponent
PrimeLock® Installation Wrench, 1" OD tubing; 1/ea
PrimeLock® Installation Wrench, 1" OD tubing; 1/ea
Catalog # MFLX30493-43
2 month warranty
PrimeLock® Installation Wrench, 1" OD tubing; 1/ea
Catalog # MFLX30493-43
2 month warranty
About this item
Easily install your High-Purity PFA Fittings.
  • Optimize performance of your PFA Flare Fittings
Replacement parts and accessories for PrimeLock® High-Purity Fittings.
  • Colour:
    Natural
  • Description:
    Installation Wrench, 1" OD tubing; 1/ea
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    PFA
  • Compression size / tube ext.Ø (port 1):
    1"
  • Warranty:
    2 months
  • Connection type:
    Flare Wrench
Frequently Bought Together