ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Elbow, Compression to Threaded, 3/8" OD x 3/8" NPT(M); 10/PK
Masterflex® Fitting, Polypropylene, Elbow, Compression to Threaded, 3/8" OD x 3/8" NPT(M); 10/PK
Catalog # MFLX06386-50
undefined
Masterflex® Fitting, Polypropylene, Elbow, Compression to Threaded, 3/8" OD x 3/8" NPT(M); 10/PK
Catalog # MFLX06386-50
About this item
Maintain a permanent connection with these adapters.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    Elbow
  • Thread size NPT(M) (port 2):
    ⅜"
  • Description:
    Compression male pipe adapter elbow, ⅜" × ⅜", polypropylene, 10 Pack
  • Seq:
    12
  • Environmentally Preferable:
  • Model:
    06386-50
  • Orifice dimensions:
    8,73 mm (0,34375")
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    Polypropylene
Frequently Bought Together