ContactUsLinkComponent
Masterflex® Fitting, Nylon, Straight, Male Luer Lock to Hose Barb Adapter, 3/32" ID; 25/PK
Masterflex® Fitting, Nylon, Straight, Male Luer Lock to Hose Barb Adapter, 3/32" ID; 25/PK
Catalog # MFLX45505-02
CE certificate
Avantor exclusive
undefined
Masterflex® Fitting, Nylon, Straight, Male Luer Lock to Hose Barb Adapter, 3/32" ID; 25/PK
Catalog # MFLX45505-02
CE certificate
Avantor exclusive
About this item
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male Luer Lock x 3/32" ID
  • Ideal for use with flexible tubing
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
  • Barb size/tubing int.Ø (port 1):
    ³⁄₃₂"
  • Connection Type:
    Male luer to barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, nylon, straight, male luer lock to hose barb adapter, ³⁄₃₂" ID, 25/PK
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    45505-02
  • Connection type (port 1):
    Male luer lock
  • No. of ports:
    2
  • Warranty:
    3 Months
  • Pk:
    25
  • Fitting material:
    Nylon
Frequently Bought Together