BarbLock Assembly Hand Tool for 1/8" tubing; 1/Ea
Catalog # MFLX30604-18
Supplier: Avantor Fluid Handling
About this item
Easily install your Barblock Fittings.
- Optimize performance of your Barblock fittings
Select a hand tool to assemble your BarbLock Retainers based on the tubing size. Universal removal tool is also available.
- Description:Assembly Hand Tool for 1/8" tubing
- Material:PVDF/stainless steel
- Environmentally Preferable:
- Pk:1
- Max. int.Ø:0,125"
Frequently Bought Together