ContactUsLinkComponent
Masterflex® Barbed to Sanitary Adapter, ¹/₄" ID to 1¹/₂", ADCF PP; 10/PK, Avantor®
Masterflex® Barbed to Sanitary Adapter, ¹/₄" ID to 1¹/₂", ADCF PP; 10/PK, Avantor®
Catalog # MFLX31515-97
CE certificate
undefined
Masterflex® Barbed to Sanitary Adapter, ¹/₄" ID to 1¹/₂", ADCF PP; 10/PK, Avantor®
Catalog # MFLX31515-97
CE certificate
About this item
Certificate of compliance included!
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilisable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow - and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilised by gamma irradiation. Material meets the requirements for USP Class VI.
Delivery: Includes certificate of compliance.
  • Barb size/tubing int.Ø (port 2):
    1/4"
  • Connection Type:
    Sanitary to Barb
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    1-1/2"
  • Description:
    Barbed to Sanitary Adapter, ¹/₄" ID to 1-1/2", ADCF PP; 10/PK, Avantor®
  • Seq:
    09
  • Environmentally Preferable:
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Pk:
    10
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    1-1/2"
Sanitary Caps
Product Information