ContactUsLinkComponent
Eppendorf Reference 2 lower part, Single-channel lower part with separate serial number, 10 µL, medium gray, variable: 0.5 µL - 10 µL, fixed: 5 µL, 10 µL 1 * 1 items
Catalog # EPPE4920602.008
Supplier:  EPPENDORF
Eppendorf Reference 2 lower part, Single-channel lower part with separate serial number, 10 µL, medium gray, variable: 0.5 µL - 10 µL, fixed: 5 µL, 10 µL 1 * 1 items
Catalog # EPPE4920602.008
Supplier:  EPPENDORF
Frequently Bought Together